Learn More
Bel-Art™ SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System
Designed for easy injection of chemicals through a gas-tight syringe for air sensitive samples without using a glove box. Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System is comprised of a 5mm end cap, 2-way valve, septum with cap, barrel coupler and a vacuum connector.
Brand: Bel-Art™ 66000-0037
Code : 03
Additional Details : Weight : 0.12920kg
Description
- The Omni-Fit Tube consists of a 507-PP tube topped by a sturdy 2 in. section of medium-walled tubing which supports the valve system
- Omni-Fit Sample tubes (507-OF-7) sold separately
- Omni-Fit valve (OFV) is a complete accessory set
- Additional septum (OF-30) are available in packages of 6
Specifications
Omni-Fit Septum | |
5.0 mm | |
OF-30 |