missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Wilmad™ Thin Wall Quartz EPR Sample Tube

Product Code. p-6974026 Shop All SP Scienceware Products
Click to view available options
Diameter (Metric) Outer:
4 mm
5 mm
Diameter (Metric) Inner:
3 mm
4 mm
Unit Size:
Each

Product Code. 15368578

Brand: Bel-Art™ 660000014

Only null left
Add to basket
Add to basket

Manufactured from naturally occurring quartz with the highest purity to minimize the background signal. Bel-Art SP Scienceware™ Thin Wall Quartz EPR Sample Tube may contain Fe2O3 which is paramagnetic (1200 ppm) and has certain impact on shimming quality depending on the magnetic field strength.

  • Tight ID and OD tolerances ensure 100% compatibility to spectrometers
  • Individually wrapped to prevent contamination
  • Recommended for experiments with small organic molecule (Molecular Weight ∼500) at ambient temperature only

Specifications

Length (Metric) 250 mm
Material Quartz
Product Type Sample Tube
Diameter (Metric) Outer 4 mm
Diameter (Metric) Inner 3 mm
Thickness (Metric) Wall 0.5 mm
Model 707-SQ-250M
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.