missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Winter Pro Gloves
Click to view available options
Size: 8
Description
- PVC glove on 100 % seamless woven cotton knitted
- Patterned finish
- The seamless woven cotton knitted brings to the glove a very good comfort
- The PVC used to make the glove insure a good flexibility during the use
- Watertight glove with woven cotton seamless liner to protect against cold, suitable for use in wet / oily environment
- The high quality PVC ensures continued outstanding quality
- The cuff ensures good forearm protection
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 374/2-2003 Chemical hazards, EN 511-2003 Cold hazards |
Exterior Finish | Patterned |
Material | PVC |
Size | 8 |
Thickness (Metric) | 1.4 mm |
Liner Material | Cotton |
Length (Metric) | 26.7 cm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'