Learn More
Tuttnauer Wire (Mesh) Basket
Stainless steel mesh wire basket for bottles and general use.
Brand:  Tuttnauer BSK830-0002
 View more versions of this product
Product Code. 17849588
  • £145.00 / Each
Explore more special offers
Description
Description
  • Stainless Steel wire basket for the sterilization of solid items, or liquid in bottles.
  • The baskets are stackable and two baskets can be placed in the autoclave chamber.
Specifications
Specifications
6.4 gal.
28 cm
14.3 in.
Autoclave Basket
Non-sterile
29 L
11.2 in.
36.2 cm
362 x 280 mm