Learn More
Tuttnauer Wire (Mesh) Basket
Stainless steel mesh wire basket for bottles and general use.
Brand: Tuttnauer BSK830-0003
This item is not returnable.
View return policy
Description
- Stainless Steel wire basket for the sterilization of solid items, or liquid in bottles.
- The baskets are stackable and two baskets can be placed in the autoclave chamber.
Specifications
5.7 gal. | |
25 cm | |
14.3 in. | |
Autoclave Basket | |
Non-sterile |
26 L | |
10 in. | |
36.2 cm | |
362 x 250 mm |