missing translation for 'onlineSavingsMsg'
Learn More

Daniels Healthcare™ WIVA Container with Solid Lid

Product Code. 15514146
Click to view available options
Capacity (Metric):
30 L
60 L
Dimensions (L x W x H):
400 x 335 x 318 mm
400 x 335 x 640 mm
Unit Size:
Each

Product Code. 15514146

Brand: Daniels Healthcare™ UKWIVB30YELSLP81

Please to purchase this item. Need a web account? Register with us today!

Edge

Remove and dispose of potentially hazardous biological materials safely. Daniels Healthcare™ WIVA Container with Solid Lid conforms to BS 7320 specifications for waste removal.

  • Manufactured from Polypropylene
  • Includes lid
  • Suitable for clinical waste removal
  • Conforms to BS7320

Specifications

Capacity (Metric) 30 L
Certifications/Compliance BS 7320
Color Yellow
Dimensions (L x W x H) 400 x 335 x 318 mm
Height (Metric) 318 mm
Length (Metric) 400 mm
Width (Metric) 335 mm
Material Polypropylene
Product Type Waste Removal Bin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.