missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Wnt Surrogate-Fc Fusion Recombinant Protein
HEK293 cells Recombinant Protein
Brand: Invitrogen™ PHG0402
This item is not returnable.
View return policy
Description
Wnt3a, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.Spezifikation
P56704 | |
Functional Assay | |
89780 | |
100 μg | |
-20°Cor to +80°C if preferred | |
1 EU/mL | |
WNT3A | |
Unconjugated | |
Liquid |
2 mg/mL | |
25 mM tris with 500 mM NaCl and no preservative; pH 8.2 | |
Wnt Surrogate-Fc Fusion | |
HEK293 cells | |
RUO | |
MGC119418; MGC119419; MGC119420; Protein Wnt-3 A; vestigial tail; vt; wingless-related MMTV integration site 3 A; wingless-type MMTV integration site family member 3 A; wingless-type MMTV integration site family, member 3 like; wingless-type MMTV integration site family, member 3 A; Wnt 3 A; Wnt family member 3 A; wnt[a]; wnt3; wnt3 l; wnt3 like; Wnt3a; Wnt-3 A; WNT3A protein; wnt3l; Zwnt[a] | |
Human | |
Recombinant | |
>95% |