Learn More
Technical Service Consultants™ Woodstick Swab
Hygiene swab with wood shaft and cotton tip in tube
Brand: Technical Service Consultants™ TS/6-H250.
Additional Details : Weight : 0.20000kg
Description
- Gamma Irradiation to achieve a SAL of 10-6
- Recommended for hygiene and general purpose sampling
Specifications
Wood | |
Sterile | |
Aseptic or one off techniques |
Cotton | |
2 Years | |
250 |