Learn More
Brady™ WorkHorse™ B-423 Permanent Polyester Labels
For use in commercial/industrial applications. Brady B-423 polyester label are for use with BMP71 printer.
Brand:  Brady™ 115027
 View more versions of this product
Product Code. 11771835
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
UL recognized, CSA accepted, AGA approved, Halogen free (DIN VDE 0472/part 815)
Acrylic, Permanent
Gloss
Thermal Transfer portable printer label
BMP™71 Printer
5000/Roll
0.5 x 0.5 in.
-70°C to +110°C
0.076 mm
12.70 x 12.70 mm
12.70 mm
Abrasion, Chemical, Corrosion, Humidity, UV and Weather-Resistant
White
Polyester (B-423)
Asset tracking, Barcoding, Component ID, Equipment ID, General purpose ID, Rating plates
1
Square
12.7 x 12.7 mm
0.003 in.
15.88 mm
12.70 mm