Learn More
Brady™ WorkHorse™ Series Thermal Transfer Printable Labels, B-488
Used to identify property from components to finished products. Brady™ WorkHorse™ Series Thermal Transfer Printable Labels are constructed of polyester and is used for thermal transfer printers.
Brand:  Brady™ 019261
 View more versions of this product
Product Code. 13049302
  • £190.00 / 1 roll
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in various sizes
Specifications
Specifications
UL Recognized; CSA Accepted
Acrylic, Permanent
Matte
Polyester (B-488)
Asset Tracking, Barcode Labeling, Circuit Board Identification - Through Hole Top, Component and Equipment Identification, General and Industrial Identification, Rating Plates
1
Rectangle
-40°C to +160°C
0.091 mm
22.86 x 22.86 mm
22.86 mm
22.86 mm
Recommended ribbon series: R4300; R6200
White
Roll
Thermal Transfer Printer Label
BBP™81 Label Printer, IP™ Series, PR Plus Series Printers
1000/Pk.
22.86 x 22.86 mm (W x H)
0.0036 in.
25.40 mm
0.9 in.
0.9 in.