Learn More
Brady™ Workstation CLP/GHS Software Download
Align your labels with CLP and GHS classifications using the easy-to-use Workstation CLP/GHS Software.
Brand: Brady™ 195526
This item is not returnable.
View return policy
Description
The Brady™ Workstation CLP/GHS Labels app makes aligning with the CLP Classification, Labeling and Packaging Regulation (1272/2008) and the Globally Harmonized System of Classification and Labeling of Chemicals (GHS) amazingly simple.
- Easy uploading and storing of Safety Data Sheet (SDS) information and label creation capabilities
- Download Brady™ Workstation at Workstation.BradyID.com
Specifications
Brady Sign and Label Maker, Multicolor and Cut Label Printer, GlobalMark 2 Industrial Label Maker | |
Windows™ 7, Windows™ 8 | |
Hazard Communication Identification | |
BWRK-CLP-DWN-EU |
Product Content Correction
Product Title
Brady™ Workstation CLP/GHS Software Download > CLP/GHS Software Download; For Use With (Application): Hazard Communication Identification; Operating System: Windows™ 7, Windows™ 8
Spot an opportunity for improvement?Share a Content Correction