missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Wortmannin, Penicillium funiculosum, 99+%

Product Code. 15416129
Click to view available options
Quantity:
10 mg
25 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibitor of phosphatidylinositol 3-kinase

Wortmannin is used as inhibitor of phosphatidylinositol 3-kinase, used cell biology reagent that has been used previously in research to inhibit DNA repair, receptor-mediated endocytosis and cell proliferation. It inhibits fMLP induced PIP3 and superoxide anion production in guinea pig neutrophils. It blocks insulin metabolic effects in rat adipocytes without affecting the insulin receptor tyrosine kinase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19545-26-7
Molecular Formula C23H24O8, C23H24O8
Synonym KY 12420

Specifications

Quantity 50 mg
Description White to Off-white
Content And Storage -30°C to -10°C
For Use With (Application) Inhibitor of phosphatidylinositol 3-kinase
Formula Weight 428.44g/mol
Sensitivity Light Sensitive
Product Type Signal Transduction Reagent
Form Powder
Beilstein 67676
InChI Key QDLHCMPXEPAAMD-QAIWCSMKSA-N
SMILES COC[C@H]1OC(=O)C2=COC3=C2[C@@]1(C)C1=C([C@@H]2CCC(=O)[C@@]2(C)C[C@H]1OC(C)=O)C3=O
IUPAC Name (1R,3R,5S,9R,18S)-18-(methoxymethyl)-1,5-dimethyl-6,11,16-trioxo-13,17-dioxapentacyclo[10.6.1.0²,¹⁰.0⁵,⁹.0¹⁵,¹⁹]nonadeca-2(10),12(19),14-trien-3-yl acetate
Molecular Weight (g/mol) 428.44
Assay ≥99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.