missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biochrom WPA Biowave 3+ CT
The WPA Biowave 3+ CT offers all the capabilities of a research grade Life Sciences UV/Vis spectrophotometer. Expect exceptionally high resolution with the 3 nm bandwidth. The 5 in. color touch screen allows users to distinguish multiple samples by color on graph overlays and to focus on areas of interest.
Brand: Biochrom 80-3007-58
This item is not returnable.
View return policy
Description
- UV/Vis Spectrophotometer with a 3 nm bandwidth
- 5 in. color touch screen with zoom and track functions makes it easy to distinguish multiple samples by color on graph overlays
- Features a USB port for data collection
- Pre-programmed life science methodologies, including nucleic acid quantification (DNA, RNA and oligonucleotides), protein assays, and cell culture density (OD600) measurements.
- Loaded with a menu of commonly available fluorescent dyes to specifically target nucleic acids and proteins
- Nucleic acid scans for purity checking
- Low volume capability for use with microvolume cuvettes
- Choose to include a built-in serial printer, Bluetooth connectivity, or both
- Lightweight and stackable for storage
- Spectrophotometer has a 2 year warranty and the Xenon flash lamp has a 3 year warranty
- IQ/OQ available on request
Specifications
15 mm | |
390 mm | |
Color Touchscreen | |
Xenon Flash | |
3 nm | |
120/240 V | |
UV/Vis Spectrophotometry | |
3.9 in. | |
Spectrophotometer, Built-in Serial Printer | |
Flat Grating | |
±1.3% or ±0.008 A (Whichever is Greater at 546 nm) | |
±0.002 A to 0.5 A (at 546 nm) | |
Round | |
±2 nm | |
6.6 lb. | |
10 in. |
15.3 in. | |
Twin CMOS Array | |
English, French, German, Spanish, Italian, Chinese, Japanese | |
WPA Biowave 3+ CT | |
UV-Vis Spectrophotometer | |
190 to 1100 nm | |
Low Volume UV Cuvettes | |
100 mm | |
Touch Screen Display (Color), USB Port, PC via PVC Software | |
±0.005 peak to peak ±0.002 RMS | |
-0.3 to 2.5 A, 0.3 to 199 %T | |
Yes | |
<0.5 %T 340 nm | |
±1 nm | |
3 kg | |
260 mm |