Learn More
Microspec™ Wrappastack™ Sleeves
For storage of prepared petri dishes.
Brand:  Microspec™ K2
 View more versions of this product
Product Code. 15772135
  • £46.65 / Pack of 200
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Professionally wrap poured plates for fridge storage or incubation
  • Allows poured plates to heat shrunk into sleeves for fridge storage
  • Keeps plates neatly stacked and steady
  • Reduces moisture loss
  • Tear strip for easy removal of plates when required
Specifications
Specifications
Polypropylene
Sleeve
180 mm
90 mm