missing translation for 'onlineSavingsMsg'
Learn More

Wright's Stain, Thermo Scientific Chemicals

£31.96 - £84.32

Chemical Identifiers

CAS 68988-92-1
Molecular Formula C36H27Br4N3O5S+2
Molecular Weight (g/mol) 933.304
MDL Number MFCD00082143
InChI Key AXIKDPDWFVPGOD-UHFFFAOYSA-O
PubChem CID 25113599
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11429016
View Documents
Thermo Scientific Alfa Aesar
A17903.14
25 g
£31.96
25g
In Stock
to see stock.
Add to basket
 
11439016
View Documents
Thermo Scientific Alfa Aesar
A17903.22
100 g
£84.32
100g
In Stock
to see stock.
Add to basket
 
Description

Description

Useful stain for blood and for bone marrow films

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

68988-92-1
933.304
AXIKDPDWFVPGOD-UHFFFAOYSA-O
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid
C36H27Br4N3O5S+2
MFCD00082143
25113599
CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O

Specifications

68988-92-1
MFCD00082143
CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.C1=CC=C(C(=C1)C2=C3C=C(C(=C(C3=[O+]C4=C(C(=C(C=C24)Br)O)Br)Br)O)Br)C(=O)O
933.304
Wright's Stain
C36H27Br4N3O5S+2
AXIKDPDWFVPGOD-UHFFFAOYSA-O
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;2-(2,4,5,7-tetrabromo-3,6-dihydroxyxanthen-10-ium-9-yl)benzoic acid
25113599
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

missing translation for 'einecsNumber' : 273-541-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.