Learn More
Cole Parmer™ Write-On Tape
Use to label/identify laboratory items/products.
Brand:  Cole Parmer™ 06530-28
 View more versions of this product
Product Code. 15366155
  • £80.57 / Pack of 24
Edge
Explore more special offers
Description
Description
  • Identify with writing and color.
  • Durable tape is waterproof, and oil and acid resistant.
  • Can be used with pencil, pen, or permanent marker.
  • Temperature resistant.
Specifications
Specifications
Assorted
24/Pk.
14 yd. x 0.5 in.
0.5 in.
24/Pk.
-23°C to +121°C
14 yd.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole Parmer™ Write-On Tape > Assorted, 1/2 in. x 14 yds., 24 Pk