missing translation for 'onlineSavingsMsg'
Learn More

Cole Parmer™ Write-On Tape

Use to label/identify laboratory items/products.

£58.10 - £80.57

Specifications

Temperature Range (Metric) -23°C to +121°C
Length (English) 14 yd.
View More Specs

Products 2
Product Code Brand Color Width (English) Price Quantity & Availability  
Product Code Brand Color Width (English) Price Quantity & Availability  
15719404
View Documents
Cole Parmer™
06530-21
White 0.5 in.
£58.10
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15366155
View Documents
Cole Parmer™
06530-28
Assorted 0.5 in.
£80.57
Pack of 24
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Identify with writing and color.
  • Durable tape is waterproof, and oil and acid resistant.
  • Can be used with pencil, pen, or permanent marker.
  • Temperature resistant.
Specifications

Specifications

-23°C to +121°C
14 yd.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole Parmer™ Write-On Tape

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.