missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex X-Fit Disposable Earplugs
26.48 GBP valid until 2024-12-31
Use promo code "23255" to get your promotional price.
Use promo code "23255" to get your promotional price.
Click to view available options
Content And Storage:
200 Pairs
300 Pairs, Refill Box
Description
- The ergonomically pre-shaped uvex x-fit disposable earplugs provide very strong insulation and are suitable for use in extremely loud environments.
- The soft foam of the earplugs provides a high level of comfort, even when worn for extended periods.
Specifications
Content And Storage | 200 Pairs |
Certifications/Compliance | EN 352-2 standard |
Cushion Material | PU Foam |
CSA Z94.2 | Class AL |
Cushion Color | Lime |
Size | Regular |
High Visibility | Yes |
Plug Material | PU Foam |
Plug Color | Lime |
Noise Reduction Rating (NRR) | 37 db |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'