Learn More
Thermo Scientific™ X-Gal
Effectively screen colonies with X-Gal, an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal, forming an intense blue precipitate.
Brand: Thermo Scientific™ R0402
83.78 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
X-Gal (5-bromo-4-chloro-3-indolyl-beta-D-galacto-pyranoside) is an inert chromogenic substrate for beta-galactosidase which hydrolyzes X-Gal into colorless galactose and 4-chloro-3-brom-indigo, forming an intense blue precipitate. Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies.
Induction of the lacZ gene with IPTG leads to the hydrolysis of X-Gal and to the development of blue colonies. X-Gal Solution is a stable, 0.22μm membrane filtered solution formulated for direct use in conjunction with IPTG for blue/white colony screening.
Specifications
Colorimetric | |
2 x 1 g | |
Beta-Gal Substrate, X-Gal |
X-Gal | |
Chemical Substrate |
Safety and Handling
- X-Gal
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Lithium chloride, (R*,R*)-1,4-dimercapto-2,3-butanediol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.