Learn More
Abeba™ X-Light Low Safety Shoe
Abeba X-Light Low Safety Shoe has a breathable inner lining with silver ions.
Brand: Abeba™ 711038-48
Additional Details : Weight : 1.80000kg
Description
- Size 35 to 48
- CE, EN ISO 20345:2011, S2, SRC
- Smooth leather, black
- Breathable lining with Silver ions
- Instep area with lacing
- Slip resistance PU outsole sole
- Steel toe cap
- Replaceable Active Comfort Insole (art. 351620)
- Antistatic
- Reflective material
Specifications
Laces | |
Leather | |
Steel |
Black | |
48 | |
CE, EN ISO 20345:2011, S2, SRC |