Learn More
Abeba™ X-Light Safety Loafer
Abeba X-Light White Safety Loafer - Smooth leather safety shoes with elastic instep.
Brand: Abeba™ 711032/39
80.21 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : Weight : 0.75000kg
Description
- CE, EN ISO 20345:2011, S2, SRC
- Smooth leather
- Black
- Steel toe cap
- Slip resistance PU outsole sole
- Replaceable Active Comfort Insole (art. 351620)
- Antistatic
- Breathable lining with Silver ions
- Instep area with elastic band
Specifications
Elastic Band | |
Leather | |
Steel |
White | |
39 | |
CE, EN ISO 20345:2011, S2, SRC |