Learn More
Corning™ X-MINI™ Pressor
Corning™'s X-MINI™ Pressor allows for the degassing of microbubble-targeted cells. It accommodates a capped 10 mL syringe. The pressure generated using the X-MINI™ Pressor is sufficient to isolate up to 1 x 107 cells, and one syringe can process up to 3 mL of cell suspension at a time.
Brand: Corning™ 6915
Additional Details : Weight : 0.79710kg
Description
Corning™'s X-SERIES™ Systems are manufactured for Corning™ by ThermoGenesis Corp. X-SERIES™, X-WASH™, X-LAB™, X-BACS™, and X-MINI™ are either registered trademarks or trademarks of ThermoGenesis Corp. in the United States and/or other countries.Specifications
Corning X-SERIES System | |
Bioprocessing Applications |