Learn More
Draeger Safety X-Pect Glasses Product Code.: 15188414
In Stock
to see stock.
Add to basket
Draeger Safety X-Pect Glasses
Product Code. 15188414
Each
Click to view available options
Quantity:
Each
Product Code. 15188414
Brand: Draeger Safety R58249
In Stock
to see stock.
Edge
Add to basket
Add to basket
Draeger Safety X-Pect Protective Glasses ensure maximum protection for eyes against UV and exposure to harsh chemicals, which sit comfortably on the wearer.
  • Designed for maximum comfort
  • Class 1: accommodates long term use
  • Durable, with a long shelf life
  • Manufactured using quality materials
  • Soft nose bridge, to accommodate extreme weather
  • Sleek, wrap around design for wider field of view
  • Suits laboratories, medical field use, maintenance and general industrial use
Specifications
Certifications/Compliance CE certified according to EN166: 2001
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Quantity Each
UV Protection UV400
Product Content Correction
Product Title
Draeger Safety X-Pect Glasses >