Learn More
Draeger Safety X-Plore 1320 FFP2 NR D V Mask
Draeger Safety X-Plore 1320 FFP2 NR D V Mask is a disposable face mask for protection against solid and liquid particles (class 2).
Brand: Draeger Safety 3951214
Code : NEW
Additional Details : Weight : 0.25200kg
Description
- Suitable for applications including the sanding and woodcutting of hardwood and softwood
- CoolSAFE filter material produces a high filter performance and low breathing resistance
- Effective nose seal absorbs sweat and ensures a tight seal around the face
- Stable, reinforced mask body offers dimensional stability for long periods of time
- Communication is still possible whilst wearing the mask
- Inner layer is hypoallergenic and is easy on the skin
Specifications
Light Blue | |
Mask | |
Valve | |
10/Pk. |