Learn More
Draeger™ X-plore™ 6530 PC Full Face Mask
Meeting the highest demands for quality and a secure fit, the X-plore™ 6530 mask body is made from EPDM making it highly resistant and skin-friendly, offering high wearing comfort.
Brand:  Draeger™ R55795
Product Code. 10310063
  • £179.00 / Each
Estimated Shipment: 28-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Safe and comfortable fit
  • Large 180° field of vision
  • Distortion-free lens
  • Efficient ventilation system ensures fog-free lens
  • 5-point harness
  • stainless steel voice diaphragm gives clear voice communication
Specifications
Specifications
CE, EN 148-1
Standard thread Rd 40 x 1/7 in.
Personal
Polycarbonate
EN 136 Class 3 (CE mark), NIOSH (PC), AS/NZS
Full Face Mask
Adjustable 5-point
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Draeger™ X-plore™ 6530 PC Full Face Mask > Full Face Mask