Learn More
Thermo Scientific™ Buffered Peptone Water (Bottled)
Description
Pre-enrich food samples prior to the isolation of Salmonella species using these ready-prepared media bottles.
Pre-enrich food samples prior to the isolation of Salmonella species with Thermo Scientific™ Oxoid™ Buffered Peptone Water ready-prepared bottled medium. Buffered Peptone Water is a non-selective pre-enrichment medium.
Low numbers and/or sub-lethally injured Salmonella cells may be present in a test food sample. Pre-enrichment in non-selective Buffered Peptone Water allows the cells time to repair and multiply before being introduced into selective culture, improving the chances of recovering Salmonella species from the sample.
- Convenient: the pre-filled bottles are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Long shelf life: up to 12 months.
- Aids recovery of damaged Salmonella cells.
The test sample is added to Buffered Peptone Water at a ratio of 1:10 and incubated at 37° ± 1°C for 18 hours and 2 hours.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2° to 25°C |
| Format | 500 mL Wide-necked Pot |
| Manufacturing Origin | United Kingdom |
| Product Type | Microbiology Media |
| Sterility | Sterile |
| Fill Weight | 225 to 229 g |
| Packaging Type | 10 wide-necked pots with screw cap closure in a box |
| Quantity | 10/Pk. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.