missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GLP-1R PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2814P-100
This item is not returnable.
View return policy
Description
GLP-1R Monoclonal specifically detects GLP-1R in Human samples. It is validated for Flow Cytometry.
Specifications
| GLP-1R | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| GLP-1 receptor, GLP1R, GLP-1R, GLP-1-R, glucagon-like peptide 1 receptor, MGC138331 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Stains human GLP-1R transfectants but not irrelevant transfectants in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 197920 | |
| Flow Cytometry 10 μL/106 cells | |
| NP_002053 | |
| GLP1R | |
| NS0 mouse myeloma cell line transfected with human GLP-1R, Arg24-Ser463, Accession No. NP_002053 | |
| 100 Tests | |
| 2740.0 | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction