Learn More
Technical Service Consultants™ Sterile Cotton Swabs
Use Technical Service Consultants™ Sterile Cotton Swabs to transport most fastidious and non-fastidious micro-organisms. They are also recommended for clinically significant samples.
Brand:  Technical Service Consultants™ TS/5-42
Code : 03
Additional Details : Weight : 1.75500kg
Product Code. 12769955
  • £282.77 / Pack of 100
In Stock
to see stock.
Description
Description
  • Extensive range of swabs available with a choice of swab shafts including white birchwood, polypropylene (flexible) and polystyrene (breakable)
  • Tips formed from pure cotton, alginate and cotton
  • Gamma irradiated to ensure a SAL of at least 10-6
Specifications
Specifications
Polystyrene
Sterile
Peel Pouch
White
1
Dacron tip
2 Years
Hygiene Sampling, difficult sampling where a durable and flexible shaft is essential
100
Swab