Learn More
X2304 Conductive Tip blackKnights 1000ul wide-bore, Tecan-Style, 96/tray, 2 trays/blister
Brand:  Ritter Plastic 49033-0006
X2304 Conductive Tip blackKnights 1000ul wide-bore, Tecan-Style, 96/tray, 2 trays/blister
Additional Details : Weight : 0.01000kg
Product Code. 16690242
  • £351.92 / Pack of 2304
Estimated Shipment: 05-08-2024