missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo X30 Technical Buffer Solution pH11, Sachets 20ml

Brand: Mettler Toledo 30111135
97.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
X30 Technical Buffer Solution pH11, Sachets 20ml
This item is not returnable.
View return policy
Description

Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Technical buffer solution pH 11.00
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Diisopropylamine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mettler Toledo X30 Technical Buffer Solution pH11, Sachets 20ml >
Spot an opportunity for improvement?Share a Content Correction