missing translation for 'onlineSavingsMsg'
Learn More
Fisherbrand™ HDPE Wide Neck 2.5L Bottle Product Code.: 10111751
In Stock
to see stock.
Add to basket

Fisherbrand™ HDPE Wide Neck 2.5L Bottle

Product Code. 10111751
2.5 L
Click to view available options
Capacity (Metric):
2.5 L

Product Code. 10111751

Brand: Fisherbrand™ FB51442

In Stock
to see stock.
Edge
Add to basket
Add to basket
Request Bulk or Custom Formats

Tested and certified to meet UN standards for the safe transportation of hazardous materials.

  • Supplied with tamper evident polypropylene cap
  • Moulded in carrying/pouring handle
  • Color: Clear
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2.5 L
Color Clear
Diameter (Metric) Outer Neck 45 mm
Material HDPE
Width (Metric) 130mm
Certifications/Compliance Tested and certified to meet UN standards for the safe transportation of hazardous materials
Height (Metric) 295 mm
Closure Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ HDPE Wide Neck 2.5L Bottle >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.