Learn More
Fisherbrand™ Swabs in Polypropylene Tubes
Use these swabs for taking samples. Fisherbrand™ Swabs in Polypropylene Tubes have a round, compact and shred resistant head.
Brand:  Fisherbrand™ 300250.2
Product Code. 11502483
  • £382.00 / Pack of 500
Edge
Description
Description
  • Sterile dry swabs supplied in shockproof round bottom polypropylene tube
  • Sterilized by ethylene oxide
  • Produced under strict ISO 9002/EN 29002 standards
  • CE marked No.98 06 0119, Class II A
Specifications
Specifications
All Swabs Have a Round, Compact and Shred Resistant Head for Sample Taking.
ISO 9002/EN 29002,CE Marked No.98 06 0119
Cotton
2.5mm
150mm
500Pack
Sterile
Wooden Shaft
Plain
Class II A
500
Tube Swab