Learn More
Tocris Bioscience™ Xanomeline oxalate
Functionally selective M1 agonist
Brand:  Tocris Bioscience™ 3569/50
576.35 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18085332
  • £734.00 / 50mg
Explore more special offers
Description
Description
Xanomeline oxalate is a functionally selective muscarinic M1 receptor agonist (EC50 values are 0.3, 5, 42, 52 and 92.5 nM at M1, M3, M5, M4 and M2 receptors respectively). Displays a complex pharmacological profile: reversible and wash-resistant binding, resulting in full agonist activity at M1; delayed wash-resistant partial agonist activity at M2; and delayed wash-resistant full agonist activity at M4. Exhibits antipsychotic activity, and improves cognitive deficits and behavioral disturbances in Alzheimer's disease and schizophrenia.
Specifications
Specifications
>98%
Xanomeline oxalate
Xanomeline oxalate
50 mg
371.46