Learn More
XBP1S-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24868-1-AP-20UL
This item is not returnable.
View return policy
Description
The XBP1S-specific antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human XBP1S-specific. This antibody recognizes human, mouse antigen. The XBP1S-specific antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
XBP1S-Specific | |
Polyclonal | |
Unconjugated | |
20 μL | |
7494 | |
IgG |
Western Blot, Immunohistochemistry | |
293 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |