missing translation for 'onlineSavingsMsg'
Learn More

Waters Corporation XBridge Prep Amide Column

XBridge BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars.

Brand:  Waters Corporation 186006609

Encompass

 View more versions of this product

Product Code. 17113802

  • £6870.00 / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • XBridge Amide columns offer you a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 11 to enable the separation of polar analytes spanning a wide range of polarity, structural moiety and pKa.
Specifications

Specifications

30 mm
6000 psi (415 Bar)
5 μm
1/Ea.
No
100 mm
Spherical
130 Å
L68
Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers