missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation XBridge Prep Amide Column
XBridge BEH Amide columns retain extremely polar compounds, including carbohydrates and sugars.
Brand: Waters Corporation 186006606

This item is not returnable.
View return policy
Description
- XBridge Amide columns offer you a chemically stable, trifunctionally bonded amide phase that is stable from pH 2 to 11 to enable the separation of polar analytes spanning a wide range of polarity, structural moiety and pKa.
Specifications
19 mm | |
6000 psi (415 Bar) | |
5 μm | |
1/Ea. | |
No |
250 mm | |
Spherical | |
130 Å | |
L68 | |
Column |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation XBridge Prep Amide Column
Spot an opportunity for improvement?Share a Content Correction