Learn More
Waters Corporation XBridge Prep HILIC Column
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds.
£2379.00 - £19875.00
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Particle Size | 5 μm |
Pore Size | 130 Å |
Quantity | 1/Ea. |
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
17162892
![]() |
Waters Corporation
186004724 |
50 mm | 19 mm |
£2379.00
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17172892
![]() |
Waters Corporation
186004725 |
100 mm | 19 mm |
£3975.00
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17182892
![]() |
Waters Corporation
186004726 |
150 mm | 19 mm |
£4756.50
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17192892
![]() |
Waters Corporation
186004727 |
50 mm | 30 mm |
£4894.50
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17102902
![]() |
Waters Corporation
186004728 |
100 mm | 30 mm |
£7227.00
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17112902
![]() |
Waters Corporation
186004729 |
150 mm | 30 mm |
£9054.00
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17122902
![]() |
Waters Corporation
186004730 |
250 mm | 19 mm |
£6100.50
Each Save Up to £79.50 » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17132902
![]() |
Waters Corporation
186004731 |
250 mm | 30 mm |
£12754.50
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17142902
![]() |
Waters Corporation
186004732 |
50 mm | 50 mm |
£9180.00
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17152902
![]() |
Waters Corporation
186004733 |
100 mm | 50 mm |
£12523.50
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17162902
![]() |
Waters Corporation
186004734 |
150 mm | 50 mm |
£15855.00
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17172902
![]() |
Waters Corporation
186004735 |
250 mm | 50 mm |
£19875.00
Each Save Up to » |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
- More rugged than silica based HILIC phases, the BEH particle's chemical stability results in long column lifetimes.
Specifications
6000 psi (415 Bar) | |
5 μm | |
1/Ea. | |
No |
Spherical | |
130 Å | |
L3 | |
Column |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation XBridge Prep HILIC Column