Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-xt Autosampler Syringes
Allows adding of internal standards without cross contamination
Brand:  Trajan Scientific and Medical™ 2926210
 View more versions of this product
Product Code. 15219263
  • £101.00 / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Available as all-metal or PTFE-tipped plunger
Specifications
Specifications
Metal (Plunger)
23 G
0.63 mm
56 mm
Autosampler Syringe
0.11 mm
10 μL
Cone