Learn More
Trajan Scientific and Medical™ XCHANGE™ CTC PAL-xt Autosampler Syringes
Allows adding of internal standards without cross contamination
Brand:  Trajan Scientific and Medical™ 2928430
 View more versions of this product
Product Code. 15279263
  • £101.00 / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Available as all-metal or PTFE-tipped plunger
Specifications
Specifications
PTFE-tipped (Plunger)
22 G
0.71 mm
LC
Autosampler Syringe
0.17 mm
50 μL