missing translation for 'onlineSavingsMsg'
Learn More

XCT 790, Tocris Bioscience™

Product Code. 18053852
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective ERRα inverse agonist

Selective ERRα inverse agonist (IC50 = ∽400 nM). Displays no antagonist activity at ERRγ or ERα at concentrations below 10 μM. Interferes with PPARγ coactivator-1α (PGC-1α)/ERRα-dependent signaling: inhibits PGC-1α induction of ERRα and MAO-B gene expression and downregulates the constitutive transcriptional activity of ERRα in numerous cell lines.

Chemical Identifiers

CAS 725247-18-7
Molecular Formula C23H13F9N4O3S
Molecular Weight (g/mol) 596.428
InChI Key HQFNFOOGGLSBBT-AWNIVKPZSA-N
Synonym 3-4-2,4-bis-trifluoromethylbenzyloxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl acrylamide, d0zb7z, xct hplc , solid, 2e-3-4-2,4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, e-3-4-2,4-bis trifluoromethyl benzyl oxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl acrylamide, e-3-4-2, 4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, e-3-4-2,4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, 3-4-2,4-bis trifluoromethyl phenyl methoxy-3-methoxyphenyl-2-cyano-n-5-trifluoromethyl-1,3,4-thiadiazol-2-yl prop-2-enamide, 3-4-2,4-bis-trifluoromethyl-benzyloxy-3-methoxy-phenyl-2-cyano-n-5-trifluoromethyl-1,3,4 thiadiazol-2-yl-acrylamide
PubChem CID 6918788
ChEBI CHEBI:79999
IUPAC Name (E)-3-[4-[[2,4-bis(trifluoromethyl)phenyl]methoxy]-3-methoxyphenyl]-2-cyano-N-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]prop-2-enamide
SMILES COC1=C(C=CC(=C1)C=C(C#N)C(=O)NC2=NN=C(S2)C(F)(F)F)OCC3=C(C=C(C=C3)C(F)(F)F)C(F)(F)F

Specifications

Quantity 50 mg
Formula Weight 596.42
Percent Purity >99%
Chemical Name or Material XCT 790
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'