missing translation for 'onlineSavingsMsg'
Learn More

Xenon difluoride, 99.5% (metals basis), Thermo Scientific Chemicals

Fluorinating agent

£143.72 - £2192.91

Chemical Identifiers

CAS 13709-36-9
Molecular Formula F2Xe
Molecular Weight (g/mol) 169.29
MDL Number MFCD00040538
InChI Key IGELFKKMDLGCJO-UHFFFAOYSA-N
Synonym xenon difluoride, xenon fluoride, xenon fluoride xef2, unii-6poj14981p, bis fluoranyl xenon, xef2, acmc-20aj9j, xenon difluoride trace metals basis
PubChem CID 83674
IUPAC Name difluoroxenon
SMILES F[Xe]F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11324707
SDS View Documents
Thermo Scientific Alfa Aesar
039739.04
2 g
£143.72
2g
Please to purchase this item. Need a web account? Register with us today!
11334707
SDS View Documents
Thermo Scientific Alfa Aesar
039739.09
10 g
£530.64
10g
Please to purchase this item. Need a web account? Register with us today!
11314707
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
039739.18
50 g
£2192.91
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used as Fluorinating agent and oxidizing agent, gaseous etchant for silicon, particularly in the production of microelectromechanical systems (MEMS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H272-H301-H330-H314-H318;
May intensify fire; oxidizer.
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P380+P375-P372-P380-P501c

H272-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 237-251-2

missing translation for 'rtecsNumber' : ZE1294116

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Xenon difluoride, 99.5% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.