missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ XL-CLASSIC-90 Safety Storage Cabinet

For the safe and approved storage of large containers of flammable hazardous substances in working areas.

Brand:  Asecos™ 3038200130386

6010.78 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 15556865

  • £6750.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ideal for large containers and drums of approx. 200 L
  • High flexibility – optional transport base for simple in-house transport
  • Safety while decanting – end-to-end earthing link at the rear panel
  • Interior space with up to 3 m earthing wire including clamp
  • Rack system with height-adjustable storage levels allows the efficient use of existing storage capabilities
  • Can be provided with bottom collecting sumps, rack systems and shelves
  • Fire resistance of 90 minutes
  • In accordance with EN 14470-1 and TRGS 510
  • User-friendly and safe – door open arrest system as standard for convenient and safe removal or storage of containers
  • No unauthorized use – doors lockable with cylinder lock
  • Ventilation – integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications

Specifications

EN 14470-1, TRGS 510
1018 x 1555 x 2224 mm
Storage of large containers of flammable hazardous substances
1000 kg
1161 kg
Steel
864 x 1386 x 1890 mm
Wing
Light gray body; light gray wing doors; 1x powder-coated sheet steel shelf, load capacity 75 kg; 1x powder-coated sheet steel transport base, height 100 mm; 1x galvanized sheet steel bottom collecting sump with grid, capacity 235 L
2
Gray
Safety storage cabinet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.