missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ XLD Agar (Dehydrated)

Product Code. p-6082048
Click to view available options
Quantity:
500 g
2.5 kg
Unit Size:
3kg
500g
This item is not returnable. View return policy

Product Code. 10517673

Brand: Thermo Scientific™ CM0469B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate Salmonella and Shigella from food and clinical specimens with this selective medium.

Isolate Salmonella and Shigella from clinical specimens and foods with selective Thermo Scientific™ Oxoid™ XLD Agar (Dehydrated). The medium relies on xylose fermentation, lysine decarboxylation and production of hydrogen sulphide for the primary differentiation of shigellae and salmonellae from non-pathogenic bacteria. The careful addition of sodium desoxycholate inhibits the growth of coliforms without decreasing the ability to support shigellae and salmonellae.

Salmonella and Shigella are major causes of bacterial enteric illness with transmission occurring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water.

Use XLD Medium for the screening of samples containing mixed flora suspected of harboring enteric pathogens, e.g., medical specimens or food products.

  • Easy-to-read: easily differentiate Salmonella (red colonies with black centers) from Shigella (red colonies) and other coliforms (yellow, opaque colonies).
  • Superior sensitivity and selectivity: exceeds that of the traditional plating media e.g., Eosin Methylene Blue and Bismuth Sulphite Agars, which tend to suppress the growth of shigellae.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description XLD Medium
Form Powder
Product Type Agar
Quantity 500 g
Yield For 9.4L medium

For Laboratory Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.