missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ XLD Agar

Product Code. p-7000002
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
XLD Agar
Manufacturing Origin:
United Kingdom
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12942588

Brand: Thermo Scientific™ PO0164A

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Isolate and identify Salmonella and Shigella spp. with a selective medium that relies on xylose fermentation, lysine decarboxylation and production of hydrogen sulphide.

Isolate and identify Salmonella and Shigella spp. with Thermo Scientific™ XLD Agar, a selective medium that relies on xylose fermentation, lysine decarboxylation and production of hydrogen sulphide for the primary differentiation of shigellae and salmonellae from non-pathogenic bacteria.

XLD Agar is a selective media for the isolation and differentiation of Salmonella, relying on xylose fermentation, lysine decarboxylation and production of hydrogen sulfide for the primary differentiation of Shigella and Salmonella spp. from non-pathogenic bacteria. The sensitivity and selectivity of XLD Agar exceeds that of traditional plating media, e.g., Eosin Methylene Blue and Bismuth Sulphite agars, which tend to suppress the growth of shigellae. The recovery of salmonellae and shigellae is not obscured by profuse growth of other species; therefore, XLD Agar is ideal for the screening of samples containing mixed flora and suspected of harbouring enteric pathogens, e.g., medical specimens or food products. XLD Agar is recommended in the ISO 6579 testing of Salmonella and is tested according to ISO 11133-2.

Salmonella and Shigella are major causes of bacterial enteric illness with transmission occuring via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water. A relatively small number of organisms ingested is capable of causing symptomatic infection. Salmonella and Shigella are aerobic, gram-negative, oxidase-negative bacilli. Some Salmonella and all Shigella are non-motile.

This media is tested according to ISO 11133-2 and is recommended in the ISO 6579 for testing of Salmonella.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Tested in accordance with CEN ISO/TS 11133-2. According ISO 6579
Color Red, clear
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Media Type XLD Agar
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.