Learn More
Thermo Scientific™ Xpect™ C. difficile Toxin A/B Test
Rapidly detect Clostridium difficile toxin A and B from human fecal samples with a simple, three-step procedure and results in 20 minutes.
Brand: Thermo Scientific™ R24640
Code : NEW
Additional Details : Weight : 0.05000kg
Description
Rapidly detect Clostridium difficile toxin A and B from human fecal specimens using Thermo Scientific™ Xpect™ C. difficile Toxin A/B Test. Combine excellent clinical performance with a simple, three-step procedure that offers accurate results within 20 minutes with less than one minute hands-on time. When you need STAT testing or 24/7 coverage, choose Xpect.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Clostridium difficile toxin A and B. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple walk-away procedure.Xpect C. difficile Toxin A/B uses antibody-coated colored particles to bind with target antigen in the diluted sample. The colored antibody/antigen complex moves along the test membrane. Immobilised antibodies specific to target antigen capture the color-labelled antigen complex at the test region, causing a clearly visible line to appear. This line will not appear in the absence of specific antigen. Immobilised, non-specific antibodies in the control region capture the color-labelled antibodies, whether antigen is present or not, resulting in a clearly visible control line.
- Clearly visible, easy-to-read results within 20 minutes
- Less than one minute hands-on time
- Integral control for added confidence
- Excellent sensitivity and specificity
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 30°C | |
Xpect C. difficile Toxin A/B Test | |
Test devices, conjugate reagent 1 (3.6mL), conjugate reagent 2 (3.0mL), specimen diluent, positive control (2.0mL), negative control (2.0mL), disposable transfer pipettes, dilution tubes, wooden applicator sticks | |
40 Tests/Kit |