missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Xpect™ Flu A & B Test

Rapidly detect Influenza A and Influenza B viral antigens from human nasal wash, nasal swab and throat swab specimens, and get results in 15 minutes.

Brand:  Thermo Scientific™ R24600

657.10 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 13225629

  • £684.00 / 20 tests

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

2°C to 8°C
Influenza
Reagent
20 Tests
Xpect Flu A and B Test
Nasopharyngeal
Test devices, specimen diluent (20mL), dilution tubes, disposable transfer pipettes, flu A+/B- control swab and flu B+/A- control swab
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number245300 Certificate TypeCertificate of Analysis Date12/03/2025 Catalog NumberR24600
Lot Number245299 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog NumberR24600
Lot Number237836 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberR24600
Lot Number203173 Certificate TypeCertificate of Analysis Date12/12/2024 Catalog NumberR24600
Lot Number187610 Certificate TypeCertificate of Analysis Date03/11/2024 Catalog NumberR24600
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Xpect™ Flu A & B Test > 20 tests

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.