Learn More
Thermo Scientific™ Xpect™ Legionella Test
Rapidly detect Legionella from human urine specimens by simply adding the urine sample to the device and then reading the results.
Brand: Thermo Scientific™ R24680
Description
Optimize your workflow by rapidly detecting Legionella pneumophila antigen directly from human urine specimens using Thermo Scientific™ Xpect™ Legionella Test. Combine excellent clinical performance with a simple, one-step procedure that offers accurate results in 45 minutes with strong positive results visible within 15 minutes.
Use this rapid, lateral flow immunoassay for the direct qualitative detection of Legionella pneumophila antigen. Enhance laboratory performance and efficiency with excellent clinical sensitivity and specificity and a simple one-step procedure.- Clearly visible, easy-to-read results in less than one hour
- Detects both L. pneumophia serogroups 1 and 6
- Less than one minute hands-on time
- Excellent sensitivity and specificity
- Convenient room-temperature or refrigerated storage
Trust the entire family of Xpect rapid tests to deliver unparalleled diagnostic performance while offering ease-of-use, convenience and time savings. Each test is an immunochromatographic assay with a simple procedure that provides rapid, cost-effective detection to aid in the diagnosis of infections.
Xpect Flu A&B
Xpect RSV
Xpect C. difficile Toxin A/B
Xpect Rotavirus
Xpect Giardia
Xpect Cryptosporidium
Xpect Giardia/Cryptosporidium
Xpect Legionella
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 25°C | |
Legionella | |
Test devices, disposable transfer pipettes, positive control (1.0mL), negative control (1.0mL) |
Xpect Legionella Test | |
Legionellosis Test | |
20 Tests/Kit |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.