missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ XPert Weigh Box

Provides a leak-tight physical barrier to protect operator from hazardous airborne particulates and powders

Brand:  Labconco™ 5220320

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11751459

  • £13240.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • One-piece molded, medium-density polyethylene shell
  • Dry powder epoxy-coated steel superstructure and interior shelf of transfer chamber
  • 0.25 in. (0.64 cm) thick laminated safety glass viewing window with powder-coated steel frame and neoprene gasket
  • Upward-pivoting inner and outer transfer chamber doors counterbalanced and equipped with quick-latches
  • Black, chemical-resistant phenolic core work surface sealed to glove box floor
  • Dual HEPA filters boast 99.99% efficiency against particulates down to 0.3 microns
  • 0.05 ppm or lower average tracer gas levels detected during perimeter scan testing surpass ASHRAE™ 110 standards
  • Testing with mass spectrometer reveals no detectable leaks > 1 x 10-3mL/sec. when tested while glove box pressurized with He at 1 in. water gauge
  • Vibration < 1.5 x 10-5 in. displacement when measured at work surface at both min and max airflow volumes
  • CE conformity marking

Includes:

  • HEPA inlet and outlet filters
  • Anti-static ionizer fan to reduce static charge
  • Balance vibration isolator

Specifications:

  • Main chamber: 0.37 m3
  • Viewing window cutout dimensions (L x W): 67.1 x 31.8 cm (26.37 x 12.5 in.)
  • Overall dimensions (L x W x H): 80 x 134 x 102 cm (31.625 x 52.75 x 40 in.)
  • Main chamber interior dimensions (D x W x H): 69.9 x 85.1 63.5 cm (27.5 x 33.5 x 25 in.)
  • Transfer chamber dimensions (nominal I.D. x L): 28 x 30 cm (11 x 12 in.)
  • Shipping weight: 127 kg (280 lb.)
Specifications

Specifications

Weigh Box
Fluorescent
4 A
31.6 in.
80 cm
80 x 134 x 102 cm
40 in.
102 cm
50 Hz
147 cm
2
2 glove ports
Medium
1 Year
159 kg
33.5 in.
76.2 cm
HEPA
Polyethylene
CE
27.5 in.
70 cm
Pharmaceutical research, nanotechnology and biochemistry
25 in.
63.5 cm
57.2 in.
76.2 cm
Pigtail
1 Ea.
230 V
350 lb.
57.2 in.
147 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers