missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Waters Corporation XSelect CSH C18 Column
XSelect CSH C18 columns can be used as general purpose columns that provide alternative selectivity compared to other C18 sorbents.
Brand: Waters Corporation 186005256
This item is not returnable.
View return policy
Description
- Charged Surface Hybrid (CSH) technology helps you overcome column related problems such as peak shape asymmetry and poor loading for basic compounds, column bleed, and slow mobile-phase re-equilibration.
Specifications
2.1 mm | |
6000 psi (415 Bar) | |
3.5 μm | |
1/Ea. | |
Yes |
100 mm | |
Spherical | |
130 Å | |
L1 | |
Column |