Learn More
Waters Corporation XSelect CSH Fluoro-Phenyl XP Column
Designed to maximize selectivity differences, the XSelect CSH Fluoro-Phenyl column provides you with maximum method development freedom, especially when using low pH mobile phases.
Brand:  Waters Corporation 176002609
Encompass
 View more versions of this product
Product Code. 17142542
  • £2998.50 / Pack of 3
Explore more special offers
Description
Description
  • The non-endcapped sorbent provides you with enhanced retention of acidic compounds.
  • Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Specifications
Specifications
3 mm
18000 psi (1240 Bar)
2.5 μm
3/Pk.
No
100 mm
Spherical
130 Å
L43
eXtended Performance (XP) Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Waters Corporation XSelect CSH Fluoro-Phenyl XP Column